Deionized Water Support for Recirculating Chillers
Actual product may vary
Thermo Scientific™
Deionized Water Support for Recirculating Chillers
Maintain the resistivity of your cooling water with Thermo Scientific™ Deionized Water Support for Recirculating Chillers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
084784ThermoFlex with DIC
611003000017System IV heat exchangers, 115 V
611006000009System III heat exchangers, 120 V
598000000010ThermoFlex with IDI
611006000006Merlin Chillers with a PD1/PD2 pump
611003000016System III heat exchangers, 115 V
Catalog number 084784
Price (INR)
-
For Use With (Equipment):
ThermoFlex with DIC
Maintain the resistivity of your cooling water with Thermo Scientific™ Deionized Water Packages.
  • Maintains a water resistivity level between 1 and 3 MOhmcm
Includes: Kits include a filter canister, DI media, plumbing, connectors, filter, and a LED indication to let you know when the media needs to be replaced. DI Cartridges include only the replacement DI media.
Specifications
For Use With (Equipment)ThermoFlex with DIC
IncludesDI cartridge for all ThermoFlex equipped with a Deluxe controller and controlled DI(DIC). Not intended for units with IDI
TypeDeionized Water Support
Unit SizeEach