Dionex™ ASE™ 150/350 Cellulose Thimbles
Actual product may vary
Dionex™ ASE™ 150/350 Cellulose Thimbles
Dionex™ ASE™ 150/350 Cellulose Thimbles
Thermo Scientific™
Dionex™ ASE™ 150/350 Cellulose Thimbles
Ensure reliable and reproducible analytical results. Use these high purity cellulose thimbles to hold solid and semisolid samples in place and prevent plugging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
08834510 mL
08834722 mL
08834934 mL
08835166 mL
088353100 mL
Catalog number 088345
Price (INR)
-
Volume (Metric):
10 mL
Ensure reliable and reproducible analytical results. Use these high purity Thermo Scientific™ Dionex™ ASE™ 350/150 Cellulose Thimbles to hold solid and semisolid samples in place and prevent plugging of your Dionex ASE 350/150 Accelerated Extraction System. Made from alpha cellulose, these thimbles have excellent mechanical strength. The thimbles' dimensions accurately match Dionex ASE 350/150 System Extraction Cells to ensure an optimum fit during accelerated solvent extraction.
Excellent Mechanical Strength• Simplifies cell cleaning when extracting sticky samples or samples that melt at high temperatures• Enables direct weighing of samples and avoids sample loss during transfer• Prevents fine particulate samples from clogging the cell
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description10 mL ASE 150/350 Cellulose Thimbles
MaterialCellulose
Quantity25
Volume (Metric)10 mL
Dimensions (D x W x H)Inner Diameter: 12 mm +0/-2.0 mm, External Height: 50 mm ±1 mm; Wall Thickness: 1.5 mm ±0.5 mm
For Use With (Equipment)Dionex ASE 150 and 350
TypeCellulose Thimbles
Unit SizePack of 25