Dionex™ IonPac™ NG1-10μm Guard Column (2 x 50 mm)
Thermo Scientific™

Dionex™ IonPac™ NG1-10μm Guard Column (2 x 50 mm)

Green features
Column Format: Guard Column; Column Phase: Reversed Phase, Ion-Pairing, and Ion Suppression
Have Questions?
Catalog number 088763
Price (INR)
-

Achieve high-efficiency separations of hydrophobic, ionizable compounds with Thermo Scientific™ Dionex™ IonPac™ NS1 10μm Analytical & Guard Columns. The columns of choice for ion-pair or ion-suppression chromatography, these polymer-based, reversed-phase columns provide excellent resolution and peak shape. Packing is a neutral, macroporous, high surface area, ethylvinylbenzene polymer cross linked with 55% divinylbenzene, making IonPac NS1 columns resistant to solvents, acids and bases while permitting the use of eluent from pH 0 to 14.

Polymer-Based Columns for Efficient Reversed-Phase Separations
  • Ideal for separation of large molecules that carry localized charges, such as surfactants
  • Can be used for traditional polymeric reversed-phase applications
  • Use with ion-pair chromatography for difficult separations
  • Available in 5 and 10μm particle sizes
Key Applications

Use IonPac NS1 columns for reliable separations of various compounds: High molecular weight aliphatic, carboxylic acids; Alkyl and aromatic sulfates and sulfonates; Quaternary ammonium ions; Water-soluble vitamins; Sulfur oxides; Metal cyanide complexes; Phenols

Specifications
Flow Rate1 mL/min
For Use With (Application)Reversed Phase, Ion-Pairing, Ion-Suppression
Green FeaturesFSC
Max. Pressure300 psi (20 bar)
Packing MaterialMacroporous Ethylvinylbenzene Polymer
Particle Size10 μm
Stationary PhaseDivinylbenzene
TypeMixed Mode
Column TypeGuard
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)50 mm
Product LineProPac
pH0 to 14
Unit SizeEach