Product Image
Thermo Scientific™

DRI™Serum Toxicology Barbiturate Assay

Identify the overdose class of drug in emergency room situations where urine samples are not readily available with this Barbiturate serum toxicology assay.
Have Questions?
Catalog NumberQuantity
091125 x 8 mL
Catalog number 0911
Price (INR)
-
Quantity:
25 x 8 mL
Identify the class of drug in emergency room situations where urine samples are not readily available with Thermo Scientific™ DRI™ Serum Toxicology Barbiturate Assay. Provided in convenient packaging with separate calibrators, these liquid, ready-to-use assays have the flexibility to run on a variety of clinical chemistry analyzers, providing objective results.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSerum Toxicology Barbiturate
Quantity25 x 8 mL
Sample TypeSerum
TypeReagent
Detectable AnalytesBarbiturates (BAR)
Unit SizeEach
Contents & Storage
2°C to 8°C