Fiberlite™ F9-4 x 1000y Fixed-Angle Rotor
Actual product may vary
Thermo Scientific™
Fiberlite™ F9-4 x 1000y Fixed-Angle Rotor
This product has been discontinued.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
096-041042Jouan KR25i Centrifuge
096-041032Beckman J2 Centrifuges
096-041041Jouan KR25i Centrifuge
096-041043Himac CR21/CR22 Centrifuges
76981Sorvall RC-6 Plus, Sorvall Evolution RC Series
Catalog number 096-041042
Price (INR)
-
For Use With (Equipment):
Jouan KR25i Centrifuge
This product has been discontinued. Please consider our recommended replacement product: Fiberlite F10-4x1000 LEX Fixed Angle Rotor.
Fiberlite rotors:
  • Lightweight carbon fiber construction reduces run-times by creating higher g-forces.
  • Corrosion-and fatigue-resistant design ensures structural integrity.
  • Rotors are backed by a 15-year warranty.
Compatible with:
Thermo Scientific Sorvall™ RC-6 Plus, Sorvall Evolution™ RC Series, Beckman™ J2 Series and Hitachi Himac CR21/CR22 Series centrifuges, Nalgene™ bottles and tubes
Recommended for:
Bacterial processing, subcellular organelles from tissue homengates, clarification of cell lysates and debris from water studies.
Note:
Performance may vary by centrifuge model.
Specifications
Angle22°
DescriptionThis product has been discontinued
For Use With (Equipment)Jouan KR25i Centrifuge
K Factor5950
Max. Tube Diameter98 mm
Max. Tube Length95 mm
Min. RCF2,268 x g
Net Weight31.5 lb. (14.29 kg)
Radius (Metric)168 mm
Radius (Metric) Min.25 mm
Capacity4 x 1000 mL
MaterialCarbon Fiber Composite
Max. RCF15,241 x g
Max. Speed9,000 rpm
TypeFixed Angle Rotor
Unit SizeEach