Fiberlite™ F14-6 x 250y Fixed Angle Rotor
Actual product may vary
Thermo Scientific™

Fiberlite™ F14-6 x 250y Fixed Angle Rotor

Process large volumes with the six-place, 250 mL fixed angle rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
096-062038Beckman J2 Series Centrifuges
096-062128Thermo Scientific Jouan KR22i Centrifuges
096-062132Hitachi Himac CR21/CR22 Series Centrifuges
096-062107Beckman Avanti J Series (except the Avanti J-HC) Centrifuges
Catalog number 096-062038
Price (INR)
-
For Use With (Equipment):
Beckman J2 Series Centrifuges

Process large volumes with the six-place, 250mL Thermo Scientific™ Fiberlite™ F14-6x250y Fixed Angle Rotor.

Fiberlite rotors:
  • Run times are reduced with higher G-forces achieved through lightweight carbon fiber rotors.
  • Corrosion-and fatigue-resistant design ensures structural integrity.
  • Rotor is backed by a 15-year warranty.

Compatible with:

Thermo Scientific Sorvall™ RC-6 Plus, Sorvall Evolution™ RC Series, Thermo Scientific Jouan KR22i, Beckman™ J2 Series, Beckman J2, Avanti Series, Hitachi™ Himac CR21/CR22 Series, Nalgene™ bottles and tubes

Recommended for:

Processing lysates, homogenates, ammonium sulphate precipitation of proteins and enzymes and plant tissue homogenates.

Performance may vary by centrifuge model.

Specifications
Angle23°
DescriptionFiberlite F14-6 x 250y Fixed Angle Rotor
For Use With (Equipment)Beckman J2 Series Centrifuges
K Factor1690
Max. Tube Diameter62 mm
Max. Tube Length135 mm
Min. RCF8,122 x g
Net Weight15.5 lb. (7.03 kg)
Radius (Metric)137 mm
Radius (Metric) Min.37 mm
BiocontainmentYes
Capacity6 x 250 mL
MaterialCarbon Fiber Composite
Max. RCF30,100 x g
Max. Speed14,000 rpm
TypeFixed Angle Rotor
Unit SizeEach