Fiberlite F5-10x1000 LEX Rotor
Fiberlite F5-10x1000 LEX Rotor
Fiberlite F5-10x1000 LEX Rotor
Fiberlite F5-10x1000 LEX Rotor
Thermo Scientific™
Fiberlite F5-10x1000 LEX Rotor
Accelerate bioprocessing with this 10-liter capacity rotor.
Have Questions?
Catalog NumberCapacity
096-10100010 x 1000 mL
Catalog number 096-101000
Price (INR)
-
Request A Quote
Capacity:
10 x 1000 mL
Accelerate bioprocessing with the 10-liter capacity of the Thermo Scientific™ Fiberlite™ F5-10x1000 LEX rotor.
Batch biprocessing with the Thermo Scientific™ Sorvall™ RC BIOS 10 centrifuge system.
High throughput reduces cycle times by spinning up to 10 x 1 liter bottles in a single spin.
Enhance productivity with Fiberlite carbon fiber rotors
  • Unequalled structural integrity:
    • Fatigue resistance eliminates the need to derate or limit the speed over the rotor lifespan
    • Corrosion-resistant design protects against ever-present hazard due to moisture, chemicals, alkaline solutions or other salts in the laboratory
  • Load tubes directly into the rotor, eliminating multi-canister assemblies, which can be misplaced or damaged
  • Secure investment, backed by a 15-year warranty
Ordering information:
Order rotor body (096-01000) for complete assembly. For refresher kit, order 021-101000; Lid assembly, order 099-101000; replacement O-rings for lids, order 022-101000 separately. Contact your sales representative for information.
Compatible with:
Thermo Scientific™ Sorvall™ RC BIOS 10, Thermo Scientific™ Nalgene™ bottles and tubes
Specifications
Angle20°
DescriptionFiberlite F5-10x1000 LEX Rotor
For Use With (Equipment)Thermo Scientific Sorvall RC BIOS 10 Large Capacity Centrifuges
K Factor6662
Radius (Metric)275 mm
Radius (Metric) Min.124 mm
Capacity10 x 1000 mL
MaterialCarbon Fiber Composite
Max. RCF9,333 x g
Max. Speed5,500 rpm
TypeFixed Angle Rotor
Unit SizeEach