Fiberlite™ F13-14 x 50cy Fixed Angle Rotor
Actual product may vary
Thermo Scientific™
Fiberlite™ F13-14 x 50cy Fixed Angle Rotor
Execute a complete workflow in a single conical tube, achieving time and cost efficiencies as well as waste reduction with this fixed angle rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
096-145007Beckman J2 Series Centrifuges
096-145014Thermo Scientific Jouan KR22i Centrifuges
096-145015Hitachi Himac CR21/CR22 Series Centrifuges
096-145008Beckman Avanti Series Centrifuges
Catalog number 096-145007
Price (INR)
-
For Use With (Equipment):
Beckman J2 Series Centrifuges
Support a complete workflow in single conical tube with the Thermo Scientific™ Fiberlite™ F13-14 x 50cy Fixed Angle Rotor, which enables time and cost efficiencies as well as waste reduction.
Features:
  • Reduce run times with higher G-forces achieved through lightweight carbon fiber rotors
  • Corrosion-and fatigue-resistant design for structural integrity
  • Backed by a 15-year warranty
  • Compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions
Compatible with:
Beckman™ J2 Series, Avanti Series (except Avanti J-HC), Thermo Scientific Sorvall™ RC 6 Plus Series, Jouan KR22i and Hitachi™ Himac™ CR21/CR22 Series centrifuges.
Recommended for:
Subcellular fractions, bacteria pelleting, lysate clarification, cell membranes, sucrose gradient isolution of large sub cellular particles and gradient separation of cells.
Note:
†Performance may vary by centrifuge model.
Specifications
Angle34°
Certifications/ComplianceBiocontainment Certified by CAMR, HPA, Porton Down, U.K.
DescriptionFiberlite F13-14 x 50cy Fixed Angle Rotor
For Use With (Equipment)Beckman J2 Series Centrifuges
K Factor916
Max. Tube Diameter29 mm
Max. Tube Length115 mm
Min. RCF15,710 x g
Net Weight15.5 lb. (7.03 kg)
Radius (Metric)153 mm
Radius (Metric) Min.83 mm
BiocontainmentYes
Capacity4 x 50 mL
MaterialCarbon Fiber Composite
Max. RCF29,000 x g
Max. Speed13,000 rpm
TypeFixed Angle Rotor
Unit SizeEach