Fiberlite™ F21-48 x 1.5 Fixed-Angle Rotor
Actual product may vary
Thermo Scientific™
Fiberlite™ F21-48 x 1.5 Fixed-Angle Rotor
Support micro-volume protocols using the Thermo Scientific™ Fiberlite™ F21-48 x 1.5 Fixed-Angle Rotor.
Have Questions?
Catalog NumberFor Use With (Equipment)
096-484025Thermo Scientific Jouan KR22i Centrifuges
Catalog number 096-484025
Price (INR)
-
For Use With (Equipment):
Thermo Scientific Jouan KR22i Centrifuges
Support micro-volume protocols using the Thermo Scientific™ Fiberlite™ F21-48 x 1.5 Fixed-Angle Rotor.
Fiberlite Rotor:
  • Reduces run times with higher G-forces achieved witj lightweight carbon fiber rotors
  • Ensures structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Compatible with:
Thermo Scientific Sorvall™ RC-6 Plus Series, Jouan KR22i series centrifuges, Thermo Scientific Nalgene™ bottles and tubes
Recommended for:
Purifying viruses, pelleting mammalian cells, subcellular organelles, clarifying cell lysates and pelleting nucleic acids.
Note:
Performance may vary by centrifuge model.
Specifications
Angle25°
DescriptionFiberlite F21-48 x 1.5 Fixed Angle Rotor
For Use With (Equipment)Thermo Scientific Jouan KR22i Centrifuges
K Factor221
Max. Tube Diameter11 mm
Max. Tube Length40 mm
Min. RCF31,610 x g
Net Weight8.4 lb. (3.81 kg)
Radius (Metric)97 mm
Radius (Metric) Min.64 mm
BiocontainmentYes
Capacity48 x 1.5 mL
MaterialCarbon Fiber Composite
Max. RCF43,500 x g
Max. Speed21,000 rpm
TypeFixed Angle Rotor
Unit SizeEach