Search
| Catalog Number | Max. Speed | Max. RCF | For Use With (Equipment) |
|---|---|---|---|
| 096062148 | 11,000 rpm | 18,566 x g | Thermo Scientific Sorvall Super T21 Centrifuge |
| 75006517 | 10,000 rpm | 15,316 x g | Thermo Scientific Sorvall Legend T Plus and Multifuge 3 Plus Centrifuges |
Increase performance with versatility, speed and a robust, corrosion-free carbon fiber design by using the Thermo Scientific™ Fiberlite™ F14-6 x 250LE Fixed Angle Rotor, which supports a range of capacities and applications.
Ordering Information:
Tube adapters are sold separately. Order O-ring Refresher Kit (021-062-153) for replacements. Contact your sales representative for information.
Compatible With:
Thermo Scientific™ Sorvall™ Super T21, Sorvall Legend™ XT, T+, Multifuge™ X3, 3+ Series, Beckman™ GS-6, Allegra™ 6, GP Series.
Recommended for:
Pelleting bacteria or yeast cells and for purifying nucleic acids and cells.
Note:
†Performance may vary by centrifuge model.