Product Image
Thermo Scientific™
DRI™ Serum Toxicology Calibrator 1
Use with serum toxicology immunoassays.
Have Questions?
Catalog NumberQuantity
09635 mL
Catalog number 0963
Price (INR)
-
Quantity:
5 mL
Use with the serum toxicology Barbiturate, serum toxicology Benzodiazepine, and Tricyclics immunoassays. The Thermo Scientific™ DRI™ Serum Toxicology Calibrator 1 is a liquid, ready-to-use calibrators conveniently packaged for use with the serum toxicology immunoassays.
  • Multianalyte calibrators
Ordering Information:
0963: 1 bottle, 5mL. Concentrations: Barbiturate 500ng/mL, Benzodiazepine 25ng/mL, Tricyclics 150ng/mL.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSerum Toxicology Calibrator 1
Quantity5 mL
TypeCalibrator
Detectable AnalytesBenzodiazepines, Benzodiazepines, Tricyclic Antidepressants, Serum Barbiturate, Serum Benzodiazepine, Serum Tricyclic
Sample TypeSerum
Unit SizeEach
Contents & Storage
2°C to 8°C