Product Image
Thermo Scientific™
DRI™ Serum Toxicology Calibrator 2
Use Thermo Scientific™ DRI™ Serum Toxicology Calibrator 2 with Serum Toxicology Barbiturate, Serum Toxicology Benzodiazepine, and Tricyclics Immunoassays.
Have Questions?
Catalog NumberQuantity
09655 mL
Catalog number 0965
Price (INR)
-
Quantity:
5 mL
Use Thermo Scientific™ DRI™ Serum Toxicology Calibrator 2 with Serum Toxicology Barbiturate, Serum Toxicology Benzodiazepine and Tricyclics Immunoassays. Liquid, ready-to-use calibrators are conveniently packaged for use with serum toxicology immunoassays.
  • Multianalyte calibrators
Ordering Alerts:
0965: 1 bottle, 5mL. Concentrations: Barbiturate 1000ng/mL, Benzodiazepine 50ng/mL and Tricyclics 300ng/mL.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSerum Toxicology Calibrator 2
Quantity5 mL
TypeCalibrator
Detectable AnalytesBenzodiazepines, Benzodiazepines, Tricyclic Antidepressants , Serum Barbiturate, Serum Benzodiazepine, Serum Tricyclic
Sample TypeSerum
Unit SizeEach
Contents & Storage
2°C to 8°C