CD Hybridoma Medium (custom product)
Product Image
Gibco™
CD Hybridoma Medium (custom product)
without iron carrierwithout L-glutamineRead more
Have Questions?
Catalog NumberQuantity
0990286DK1000 mL
Catalog number 0990286DK
Price (INR)
-
Quantity:
1000 mL
without iron carrier
without L-glutamine
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHybridomas
Endotoxin Level≤10 EU/mL, ≤ 10 EU/mL
Product TypeHybridoma Medium
Quantity1000 mL
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free, Chemically-defined, Protein-free
Serum LevelSerum-free
Without AdditivesNo Glutamine, No Phenol Red
Unit Size1,000 mL
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.