CEDIA™ Therapeutic Drug Monitoring (TDM) Assays
CEDIA™ Therapeutic Drug Monitoring (TDM) Assays
Actual product may vary
CEDIA™ Therapeutic Drug Monitoring (TDM) Assays
CEDIA™ Therapeutic Drug Monitoring (TDM) Assays
CEDIA™ Therapeutic Drug Monitoring (TDM) Assays
Thermo Scientific™
CEDIA™ Therapeutic Drug Monitoring (TDM) Assays
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
100002TS
also known as 100002
Phenytoin II Assay
100016Gentamicin II Assay
100015NAPA Assay
100003TS
also known as 100003
Phenobarbital II Assay
100014Procainamide Assay
Catalog number 100002TS
also known as 100002
Price (INR)
-
Request A Quote
Description:
Phenytoin II Assay
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays. Manufactured since 1986, they provide an excellent range of Thermo Scientific™ DRI™ TDM Assays as well as CEDIA TDM Assays and Calibrators.
  • Wide dynamic measuring range of analytes as low as 0.15ng/mL for Digoxin
  • Extended linearity for analytes like Cyclosprine at 25ng/mL to 2000ng/mL range
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionPhenytoin II Assay
Quantity17 x 17 mL
Sample TypePlasma, Serum
TypeReagent
Detectable AnalytesPhenytoin
Unit SizeEach
Contents & Storage
2°C to 8°C