Product Image
Invitrogen™

Attune™ NxT Shutdown Bottle

This is a 175 mL replacement bottle for shutdown solution for the Attune™ NxT Acoustic Focusing Cytometer.
Have Questions?
Catalog NumberQuantity
1000221541 Bottle
Catalog number 100022154
Price (INR)
39,420.00
Each
Quantity:
1 Bottle
Price (INR)
39,420.00
Each

This is a 175 mL replacement bottle for shutdown solution for the Attune™ NxT Acoustic Focusing Cytometer.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune
Quantity1 Bottle
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeShutdown Bottle
Unit SizeEach
Contents & Storage
Store at room temperature.