Product Image
Invitrogen™

Attune™ NxT Focusing Fluid Filter

This 0.2 micron filter prevents contaminants from getting into the focusing fluid reservior of the Attune™ NxT Acoustic Focusing Cytometer.
Have Questions?
Catalog NumberQuantity
1000225871 Filter
Catalog number 100022587
Price (INR)
12,743.00
Each
Quantity:
1 Filter
Price (INR)
12,743.00
Each

This 0.2 micron filter prevents contaminants from getting into the focusing fluid reservior of the Attune™ NxT Acoustic Focusing Cytometer. Filters are recommended for replacement once every three months, unless performing small particle detection when once-a-month replacement is recommended.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune
Quantity1 Filter
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeFocusing Fluid Filter
Unit SizeEach
Contents & Storage
Store at room temperature.