Product Image
Invitrogen™

Attune™ NxT Filter, 413LP

This is a replacement 413 long pass filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.Read more
Have Questions?
Catalog NumberQuantity
1000227521 filter
Catalog number 100022752
Price (INR)
25,596.00
Each
Quantity:
1 filter
Price (INR)
25,596.00
Each
This is a replacement 413 long pass filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune
Quantity1 filter
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeFilter
Unit SizeEach
Contents & Storage
Store at room temperature.