Product Image
Invitrogen™

Attune™ NxT OD2-488/10 Filter

This is a replacement OD2-488/10 filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.Read more
Have Questions?
Catalog NumberQuantity
1000227681 filter
Catalog number 100022768
Price (INR)
22,949.00
Online offer
Ends: 31-Mar-2026
38,248.00
Save 15,299.00 (40%)
Each
Quantity:
1 filter
Price (INR)
22,949.00
Online offer
Ends: 31-Mar-2026
38,248.00
Save 15,299.00 (40%)
Each
This is a replacement OD2-488/10 filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune
Quantity1 filter
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeFilter
Unit SizeEach
Contents & Storage
Store at room temperature.