CEDIA™ Heroin Metabolite Drugs of Abuse Calibrators
Actual product may vary
Thermo Scientific™

CEDIA™ Heroin Metabolite Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Heroin Metabolite Drugs of Abuse Calibrators.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
100031TS1
also known as 100031
10 ng/mL
100034TS
also known as 100034
20 ng/mL
Catalog number 100031TS1
also known as 100031
Price (INR)
-
Concentration:
10 ng/mL

Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Heroin Metabolite Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration10 ng/mL
DescriptionCEDIA; 10ng/mL; 5mL
Packaging1 Bottle
Quantity1 x 5 mL
TypeCalibrator
Detectable Analytes6-Acetylmorphine
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C