CytKick™ Max Extended Cooling Adaptor
Invitrogen™
CytKick™ Max Extended Cooling Adaptor
The CytKick Max Extended Cooling Adaptor is designed for use with the CytKick Max Autosampler and the Attune Acoustic FocusingRead more
Have Questions?
Catalog NumberQuantity
1000634821 adapter
Catalog number 100063482
Price (INR)
199,654.00
Each
Quantity:
1 adapter
Price (INR)
199,654.00
Each
The CytKick Max Extended Cooling Adaptor is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer. This cooling block maintains samples at 2-15°C for 95-100 minutes while they are being analyzed on the instrument. The cooling adaptor is brought to temperature by placing it in a refrigerator overnight or in a freezer for a minimum of two hours prior to use. If this longer cooling time is not required, please see the CytKick Max Universal Cooling Adaptor, which cools samples for ∼65 minutes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineCytKick
Quantity1 adapter
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeCooling Adapter
Unit SizeEach