CytKick™ Max Tube Rack
Invitrogen™
CytKick™ Max Tube Rack
The CytKick Max Tube Rack is designed to for use with the CytKick Max Autosampler and the Attune Acoustic FocusingRead more
Have Questions?
Catalog NumberQuantity
1000642411 Rack
Catalog number 100064241
Price (INR)
75,029.00
Each
Quantity:
1 Rack
Price (INR)
75,029.00
Each
The CytKick Max Tube Rack is designed to for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer. It allows sample uptake from 24 1.5- and 2.0-mL Eppendorf tubes without the need to remove the caps. For a cooling version of this rack, please see the CytKick Max Tube Cooling Rack.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineCytKick
Quantity1 Rack
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeTube Rack
Unit SizeEach