MGC DAU Control Sets
Actual product may vary
Thermo Scientific™
MGC DAU Control Sets
Monitor Thermo Scientific™ CEDIA™ and Thermo Scientific™ DRI™ drugs of abuse testing assay performance with Thermo Scientific MGC DAU Control Sets.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
100069Optional DAU Control Set
100200Primary DAU Control Set
100201Clinical DAU Control Set
100202Select DAU Control Set
1815440Specialty DAU Control Set
Catalog number 100069
Price (INR)
-
Type:
Optional DAU Control Set
Monitor Thermo Scientific™ CEDIA™ and Thermo Scientific™ DRI™ drugs of abuse testing assay performance with Thermo Scientific MGC DAU Control Sets. These controls contain high and low level controls at drug concentrations 25% below and above screening assay cutoffs to ensure quality laboratory results.
Key Features
  • Multi-constituent
  • For use in the validation of drug of abuse assays in human urine
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
IncludesAmphetamine: 225ng/mL and 375ng/mL; Barbiturate: 150ng/mL and 250ng/mL; Benzodiazepine: 150ng/mL and 250ng/mL; Cocaine: 225ng/mL and 375ng/mL; Methadone Metabolite: 75ng/mL and 125ng/mL; and Opiate 300: 225ng/mL and 375ng/mL
Quantity2 x 5 mL
TypeOptional DAU Control Set
Detectable AnalytesAmphetamine, Secobarbital, Nitrazepam, Benzoylecgonine, Benzoylecgonine, d-Methamphetamine, EDDP, Methadone Metabolite (EDDP), Morphine, Nitrazepam, Opiate 300 (Morphine), Secobarbital
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C