Product Image
Thermo Scientific™

DRI™ Ecstasy Drugs of Abuse Assays

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1000761 x 500 mL
100146813 x 18 mL
Catalog number 100076
Price (INR)
-
Quantity:
1 x 500 mL
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • One of the largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Select DAU Control Set
DescriptionDRI Ecstasy Drugs of Abuse Assays
Quantity1 x 500 mL
Sample TypeUrine
TypeReagent
Detectable AnalytesMethylenedioxyamphetamine (MDA), Methylenedioxy-N-ethylamphetamine (MDEA), Methylenedioxymethamphetamine (MDMA)
DoA CalibratorsEcstasy Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C