Product Image
Thermo Scientific™

MGC 240 Benchtop Analyzer Consumables

Ensure uninterrupted performance of the Thermo Scientific™ MGC 240 Benchtop Analyzer with a good supply of Thermo Scientific MGC 240 Benchtop Analyzer Consumables.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
10007626Reagent Bottle, 40 mL
10007624Cuvette
10007625Reagent Bottle, 60 mL
10007627Reagent Bottle, 20 mL
100262TS
also known as 100262
Acid Washing Solution
Catalog number 10007626
Price (INR)
-
Type:
Reagent Bottle, 40 mL

Ensure uninterrupted performance of the Thermo Scientific™ MGC 240 Benchtop Analyzer with a good supply of Thermo Scientific MGC 240 Benchtop Analyzer Consumables.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Capacity (Metric)40 mL
DescriptionWith Cap
Quantity20/Pk.
TypeReagent Bottle, 40 mL
Unit SizeEach