CytKick™ Max Universal Cooling Adaptor
Invitrogen™
CytKick™ Max Universal Cooling Adaptor
The CytKick Max Universal Cooling Adaptor is designed for use with the CytKick Max Autosampler and the Attune Acoustic FocusingRead more
Have Questions?
Catalog NumberQuantity
1000814921 Adapter
Catalog number 100081492
Price (INR)
202,197.00
Each
Quantity:
1 Adapter
Price (INR)
202,197.00
Each
The CytKick Max Universal Cooling Adaptor is designed for use with the CytKick Max Autosampler and the Attune Acoustic Focusing Cytometer. This cooling block maintains samples at 2-15°C for ∼65 minutes while they are being analyzed on the instrument. The cooling adaptor is brought to temperature by placing it in a refrigerator overnight or in a freezer for a minimum of two hours prior to use. For a longer cooling time (95-100 minutes), please see the CytKick Max Extended Cooling Adaptor.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineCytKick
Quantity1 Adapter
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeCooling Adapter
Unit SizeEach