Product Image
Thermo Scientific™

CEDIA™ Benzodiazepine Assays

Utilize the globally valued, highly accurate, and precise Thermo Scientific™ CEDIA™ Benzodiazepine Assay for very stable shelf life and lot-to-lot dependability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1000941 x 65 mL, MCC
1000853 x 17 mL, TSC
100164093 x 17 mL, Indiko
17755611 x 495 mL, LC
Catalog number 100094
Price (INR)
-
Quantity:
1 x 65 mL, MCC

Utilize the globally valued, highly accurate, and precise Thermo Scientific™ CEDIA™ Benzodiazepine Assay for very stable shelf life and lot-to-lot dependability. Applications are available for an array of clinical chemistry analyzers. The β-Glucuronidase Enzyme, when used with this assay, offers enhanced sensitivity of parent drug by hydrolyzing the glucuronide metabolites. The Thermo Scientific CEDIA panel of drugs of abuse assays is ever expanding to address emerging drug trends and testing needs.

  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Benzodiazepine Assays
Quantity1 x 65 mL, MCC
Sample TypeUrine
TypeReagent
Detectable AnalytesNitrazepam
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C