Product Image
Thermo Scientific™

CEDIA™ Methadone Metabolite Drugs of Abuse Assays

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Methadone Metabolite Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100096MCC 65 mL
100087TSTSC 3 x 17 mL
1868217LC 495 mL
Catalog number 100096
Price (INR)
-
Quantity:
MCC 65 mL
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Methadone Metabolite Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
QuantityMCC 65 mL
Sample TypeUrine
TypeMethadone Metabolite Assay
Detectable AnalytesMethadone Metabolite (EDDP)
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C