Product Image
Thermo Scientific™
CEDIA™ Methadone Drugs of Abuse Assays
Utilize the globally-valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA Methadone Drugs of Abuse Assays
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100097TS
also known as 100097
1 x 65 mL, MCC
1000883 x 17 mL, TSC
100164253 x 17 mL, Indiko
17309161 x 495 mL, LC
Catalog number 100097TS
also known as 100097
Price (INR)
-
Quantity:
1 x 65 mL, MCC
Utilize the globally-valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Methadone Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Methadone Drugs of Abuse Assays
Quantity1 x 65 mL, MCC
Sample TypeUrine
TypeReagent
Detectable AnalytesMethadone (MTD)
DoA CalibratorsPPX/Methadone Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C