Product Image
Thermo Scientific™
DRI™ General Oxidant-Detect™ Specimen Validity Test Calibrators
Ensure accuracy in testing for specimen adulterants with the liquid, ready-to-use Thermo Scientific™ DRI™ General Oxidant-Detect™ Specimen Validity Test Calibrators.
Have Questions?
Catalog NumberQuantity
100099712 x 25 mL
Catalog number 10009971
Price (INR)
-
Quantity:
2 x 25 mL
Ensure accuracy in testing for specimen adulterants with the liquid, ready-to-use Thermo Scientific™ DRI™ General Oxidant-Detect™ Specimen Validity Test Calibrators, for use on a variety of general chemistry analyzers.
The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution. All of our urine adulteration assays are liquid, ready-to-use and can be performed on a variety of general chemistry analyzers. The adulteration assays provide a complete and automated way to test for sample integrity and adulteration.
Features:
  • Most extensive menu of tests available
  • Offered in convenient sizes
  • Liquid, ready-to-use
  • Can be used on a variety of general chemistry analyzers
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration0, 200 μg/mL
DescriptionDRI™ General Oxidant Specimen Validity Test Calibrator Set
Quantity2 x 25 mL
TypeCalibrator
Detectable AnalytesGeneral Oxidants
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C