Product Image
Thermo Scientific™
DRI™ General Oxidant-Detect™ Specimen Validity Test Controls
Ensure accuracy in testing for specimen adulterants with the liquid, ready-to-use Thermo Scientific™ DRI™ General Oxidant-Detect™ Specimen Validity Test Controls.
Have Questions?
Catalog NumberQuantity
100099722 x 25 mL
Catalog number 10009972
Price (INR)
-
Quantity:
2 x 25 mL
Ensure accuracy in testing for specimen adulterants with the liquid, ready-to-use Thermo Scientific™ DRI™ General Oxidant-Detect™ Specimen Validity Test Controls, for use on a variety of general chemistry analyzers.
The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution. All of our urine adulteration assays are liquid, ready-to-use and can be performed on a variety of general chemistry analyzers. The adulteration assays provide a complete and automated way to test for sample integrity and adulteration.
  • One of the most extensive menus of tests available
  • Associated calibrators offered in convenient sizes
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ConcentrationNegative (100 μg/mL) and Positive (300 μg/mL)
DescriptionDRI™ General Oxidant Specimen Validity Test Control Set
Quantity2 x 25 mL
TypeControl
Detectable AnalytesNitrite
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C