Attune™ Xenith™ SIP Shield
Invitrogen™
Attune™ Xenith™ SIP Shield
The Attune Xenith SIP Shield is a key part of instrument safety and is available as a replacement should any damage occur to the existing part provided with the instrument.
Have Questions?
Catalog NumberQuantity
100101323CS1 Ea.
Catalog number 100101323CS
Price (INR)
3,858.00
Online offer
Ends: 30-Sep-2026
5,143.00
Save 1,285.00 (25%)
Each
Quantity:
1 Ea.
Price (INR)
3,858.00
Online offer
Ends: 30-Sep-2026
5,143.00
Save 1,285.00 (25%)
Each
The Attune Xenith SIP Shield is for use with the Attune Xenith Flow Cytometer and is a key part of instrument safety, providing a barrier between the user and the sample during acquisition of data. This replacement part is available should any damage occur to the existing part provided with the instrument.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Ea.
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeSIP Shield
Unit SizeEach