CEDIA™ Heroin Metabolite (6-Acetylmorphine) Drugs of Abuse Assays
Actual product may vary
Thermo Scientific™
CEDIA™ Heroin Metabolite (6-Acetylmorphine) Drugs of Abuse Assays
Utilize the valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Heroin Metabolite (6-Acetylmorphine) Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100108TS
also known as 100108
1 x 65 mL, MCC
1001073 x 17 mL, TSC
100152133 x 17 mL, Indiko
1001861 x 495 mL, LC
Catalog number 100108TS
also known as 100108
Price (INR)
-
Quantity:
1 x 65 mL, MCC
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Heroin Metabolite (6-Acetylmorphine) Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Select DAU Control Set
DescriptionCEDIA Heroin Metabolite (6-Acetylmorphine) Drugs of Abuse Assays
Quantity1 x 65 mL, MCC
Sample TypeUrine
TypeReagent
Detectable Analytes6-Acetylmorphine, Heroin Metabolite (6-Acetylmorphine)
DoA CalibratorsHeroin Metabolite Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C