Attune™ Xenith™ 200-nm Primary Focus Fluid Filter
Invitrogen™
Attune™ Xenith™ 200-nm Primary Focus Fluid Filter
This focusing fluid filter reduces background in your Attune Xenith Flow Cytometer by filtering contaminants over 200nm in size out of the focusing fluid as it enters the instrument.
Have Questions?
Catalog NumberQuantity
100114360S1 Ea.
Catalog number 100114360S
Price (INR)
18,606.00
Online offer
Ends: 31-Mar-2026
31,009.00
Save 12,403.00 (40%)
Each
Quantity:
1 Ea.
Price (INR)
18,606.00
Online offer
Ends: 31-Mar-2026
31,009.00
Save 12,403.00 (40%)
Each
The Attune Xenith 200-nm Primary Focus Fluid Filter is designed for use with the Attune Xenith Flow Cytometer. This filter prevents contaminants from entering the focusing fluid reservoir of the instrument. It is recommended that filters are replaced every 6&ndash:12 months, depending on usage.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Ea.
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeFilter
Unit SizeEach