Product Image
Thermo Scientific™
DRI™ Methadone Metabolite Drugs of Abuse Assay
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of DRI Methadone Metabolite Drugs of Abuse Assay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100115TS
also known as 100115
1 x 100 mL
1001161 x 500 mL
Catalog number 100115TS
also known as 100115
Price (INR)
-
Quantity:
1 x 100 mL
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Methadone Metabolite Drugs of Abuse Assay, which offers applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Primary DAU Control Set
DescriptionDRI Methadone Metabolite Drugs of Abuse Assay
Quantity1 x 100 mL
Sample TypeUrine
TypeReagent
Detectable AnalytesMethadone Metabolite (EDDP)
DoA CalibratorsMethadone Metabolite Urine Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C