Attune™ Xenith™ 405 nm Laser Line Filters
Invitrogen™

Attune™ Xenith™ 405 nm Laser Line Filters

These replacement optical filters are for those channels available off the 405 nm laser of the Attune Xenith Flow Cytometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeExcitation/Emission
100117816CSEmission Filter434/17
100117813CSEmission Filter575/15
100117814CSEmission Filter507/19
100117815CSEmission Filter549/15
100117817CSEmission Filter420/10
100117818CSEmission Filter455/14
100117819CSEmission Filter473/15
100117820CSEmission Filter747/33
100117821CSEmission Filter770/LP
100117822CSEmission Filter710/20
100117823CSEmission Filter661/20
100117824CSEmission Filter615/24
100117864CSDichroic Longpass Filter530DLP
100117865CSDichroic Shortpass Filter560DSP
100117866CSDichroic Longpass Filter427DLP
100117867CSDichroic Shortpass Filter593DSP
100117868CSDichroic Shortpass Filter488DSP
100117869CSDichroic Longpass Filter445DLP
100117870CSDichroic Shortpass Filter465DSP
100117871CSDichroic Shortpass Filter725DSP
100117872CSMirrorNot Available
100117873CSDichroic Longpass Filter640DLP
100117874CSDichroic Longpass Filter770DLP
100117875CSDichroic Longpass Filter685DLP
Catalog number 100117816CS
Price (INR)
13,203.00
Online offer
Ends: 30-Sep-2026
17,604.00
Save 4,401.00 (25%)
Each
Product Type:
Emission Filter
Excitation/Emission:
434/17
Price (INR)
13,203.00
Online offer
Ends: 30-Sep-2026
17,604.00
Save 4,401.00 (25%)
Each
The Attune Xenith Flow Cytometer is designed with a flexible optical deck and the ability to run both spectral unmixing and conventional compensation assays on one system. Part of this flexibility is delivered through user-changeable optical filters. These optical filters are for the channels available off the 405 nm laser.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorViolet
Excitation/Emission434/17
Quantity1 Ea.
Wavelength405 nm
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeEmission Filter
Unit SizeEach