Attune™ Xenith™ 561 nm Laser Line Filters
Invitrogen™

Attune™ Xenith™ 561 nm Laser Line Filters

These replacement optical filters are for those channels available off the 561 nm laser of the Attune Xenith Flow Cytometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeExcitation/Emission
100117881CSMirrorNot Available
100117825CSEmission Filter685/15
100117826CSEmission Filter720/24
100117827CSEmission Filter700/13
100117828CSEmission Filter810/25
100117829CSEmission Filter760/50
100117830CSEmission Filter860/LP
100117831CSEmission Filter844/25
100117832CSEmission Filter575/15
100117833CSEmission Filter589/15
100117834CSEmission Filter622/15
100117835CSEmission Filter605/15
100117836CSEmission Filter661/20
100117876CSDichroic Longpass Filter613DLP
100117877CSDichroic Shortpass Filter640DSP
100117878CSDichroic Longpass Filter597DLP
100117879CSDichroic Longpass Filter675DLP
100117880CSDichroic Longpass Filter582DLP
100117882CSDichroic Longpass Filter735DLP
100117883CSDichroic Longpass Filter692DLP
100117884CSDichroic Shortpass Filter707DSP
100117885CSDichroic Longpass Filter828DLP
100117886CSDichroic Longpass Filter785DLP
100117887CSDichroic Longpass Filter860DLP
Catalog number 100117881CS
Price (INR)
13,203.00
Online offer
Ends: 30-Sep-2026
17,604.00
Save 4,401.00 (25%)
Each
Product Type:
Mirror
Excitation/Emission:
Not Available
Price (INR)
13,203.00
Online offer
Ends: 30-Sep-2026
17,604.00
Save 4,401.00 (25%)
Each
The Attune Xenith Flow Cytometer is designed with a flexible optical deck and the ability to run both spectral unmixing and conventional compensation assays on one system. Part of this flexibility is delivered through user-changeable optical filters. These optical filters are for the channels available off the 561 nm laser.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorYellow
Excitation/EmissionNot Available
Quantity1 Ea.
Wavelength561 nm
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeMirror
Unit SizeEach