Attune™ Xenith™ 488 nm Laser Line Filters
Invitrogen™

Attune™ Xenith™ 488 nm Laser Line Filters

These replacement optical filters are for those channels available off the 488 nm laser of the Attune Xenith Flow Cytometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeExcitation/Emission
100117893CSDichroic Longpass Filter600DLP
100117837CSEmission Filter549/15
100117838CSEmission Filter520/40
100117839CSEmission Filter583/30
100117840CSEmission Filter615/24
100117841CSEmission Filter750/LP
100117842CSEmission Filter720/24
100117843CSEmission Filter670/30
100117888CSDichroic Shortpass Filter640DSP
100117889CSDichroic Shortpass Filter560DSP
100117890CSDichroic Longpass Filter540DLP
100117891CSDichroic Longpass Filter685DLP
100117892CSDichroic Longpass Filter750DLP
Catalog number 100117893CS
Price (INR)
13,203.00
線上優惠
Ends: 30-Sep-2026
17,604.00
Save 4,401.00 (25%)
Each
Product Type:
Dichroic Longpass Filter
Excitation/Emission:
600DLP
Price (INR)
13,203.00
線上優惠
Ends: 30-Sep-2026
17,604.00
Save 4,401.00 (25%)
Each
The Attune Xenith Flow Cytometer is designed with a flexible optical deck and the ability to run both spectral unmixing and conventional compensation assays on one system. Part of this flexibility is delivered through user-changeable optical filters. These optical filters are for the channels available off the 488 nm laser.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorBlue
Excitation/Emission600DLP
Quantity1 Ea.
Wavelength488 nm
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeDichroic Longpass Filter
Unit SizeEach