Attune™ Xenith™ 637 nm Laser Line Filters
Invitrogen™

Attune™ Xenith™ 637 nm Laser Line Filters

These replacement optical filters are for those channels available off the 637 nm laser of the Attune Xenith Flow Cytometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeExcitation/Emission
100117898CSDichroic Longpass Filter685DLP
100117894CSDichroic Longpass Filter785DLP
100117895CSDichroic Longpass Filter735DLP
100117896CSMirrorNot Available
100117897CSDichroic Shortpass Filter707DSP
Catalog number 100117898CS
Price (INR)
13,203.00
線上優惠
Ends: 30-Sep-2026
17,604.00
Save 4,401.00 (25%)
Each
Product Type:
Dichroic Longpass Filter
Excitation/Emission:
685DLP
Price (INR)
13,203.00
線上優惠
Ends: 30-Sep-2026
17,604.00
Save 4,401.00 (25%)
Each
The Attune Xenith Flow Cytometer is designed with a flexible optical deck and the ability to run both spectral unmixing and conventional compensation assays on one system. Part of this flexibility is delivered through user-changeable optical filters. These optical filters are for the channels available off the 637 nm laser.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorRed
Excitation/Emission685DLP
Quantity1 Ea.
Wavelength637 nm
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeDichroic Longpass Filter
Unit SizeEach