Attune™ Xenith™ Fluidics Umbilical Tubing
Invitrogen™
Attune™ Xenith™ Fluidics Umbilical Tubing
These parts are replacements for components within the umbilical tubing that carries fluid between the fluid cart and the Attune Xenith Flow Cytometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
100118057CSUmbilical Waste Line
100125295CSBleach Tubing Quick Connect
100098435CSGround Strap
100098432CSUmbilical Electrical Cable
100120469CSUmbilical Sleeve
100118058CSUmbilical Shut Down Line
100118059CSUmbilical Wash Line
100118060CSUmbilical Bleach Line
100118061CSUmbilical Sheath Line
Catalog number 100118057CS
Price (INR)
5,193.00
Online offer
Ends: 30-Sep-2026
6,923.00
Save 1,730.00 (25%)
Each
Product Type:
Umbilical Waste Line
Price (INR)
5,193.00
Online offer
Ends: 30-Sep-2026
6,923.00
Save 1,730.00 (25%)
Each
The Invitrogen Attune Xenith Flow Cytometer is a spectrally enabled system that provides clog-resistant performance with acoustic-assisted hydrodynamic focusing. The system has an external fluid cart that holds all the fluids required for everyday use of the system. These parts are replacements of components within the umbilical tubing that carries fluidics between the cart and the instrument.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Ea.
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeUmbilical Waste Line
Unit SizeEach