Product Image
Gibco™

Medium 199, Hanks' Balanced Salts, powder

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
10012011
also known as 10012-011
1 L
10012037
also known as 10012-037
10 L
Catalog number 10012011
also known as 10012-011
Price (INR)
-
Quantity:
1 L

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This Medium 199 is modified as follows:
WithWithout
• Hanks' salts• Sodium Bicarbonate
• L-glutamine 
• HEPES 
• Phenol Red 


The complete formulation is available.

Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. This Medium 199 formulation contains Hanks' salts for use without CO2.

Medium 199 contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). This Medium 199 formulation does not require a CO2 environment to maintain physiological pH.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Specifications
Cell TypeMammalian Cells
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeMedium 199
Quantity1 L
Shelf Life36 Months
Shipping ConditionRoom Temperature
Culture TypeMammalian Cell Culture
FormPowder
With AdditivesGlutamine, HEPES, Phenol Red, Ribonucleosides, Deoxyribonucleosides, Sodium Bicarbonate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

Frequently asked questions (FAQs)

What is the shelf life of my powdered media once I reconstitute it?

Expiration date of most Gibco reconstituted dry format media (AGT or DPM) has not been established; end users should assess performance and stability of this reconstituted media in a system that is relevant to their process. A dry format product (either DPM or AGT) will age upon storage and while we may not be able to detect which component(s) degrade since we can't measure every component, that doesn't mean that the reconstituted liquid will have the same stability as a fresh liquid made by direct weigh methods.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.