Attune™ Xenith™ Bottle Caps
Invitrogen™
Attune™ Xenith™ Bottle Caps
These parts are replacement bottle caps for the containers on the fluidics cart of the Attune Xenith Flow Cytometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
100120481CSDI Water Bottle Cap
100120476CSFocus Cube Cap
100120477CSWaste Cube Cap
100120478CSWash Bottle Cap
100120479CSBleach Bottle Cap
100120480CSShutdown Bottle Cap
Catalog number 100120481CS
Price (INR)
11,690.00
Precio exclusivo compra web
Ends: 30-Sep-2026
19,483.00
Save 7,793.00 (40%)
Each
Product Type:
DI Water Bottle Cap
Price (INR)
11,690.00
Precio exclusivo compra web
Ends: 30-Sep-2026
19,483.00
Save 7,793.00 (40%)
Each
The Invitrogen Attune Xenith Flow Cytometer has an external fluid cart that holds the fluids required for everyday use of the system. These parts are replacement bottle caps for the cart’s fluidics containers, offered here in the event that damage occurs to the bottle caps provided with the instrument.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Ea.
For Use With (Equipment)Attune Xenith Flow Cytometer
Product TypeDI Water Bottle Cap
Unit SizeEach