Product Image
Thermo Scientific™

DRI™ Methadone Metabolite Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Methadone Metabolite Drugs of Abuse Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
100120TS
also known as 100120
1000 ng/mL
100117150 ng/mL
100118TS300 ng/mL
1001222000 ng/mL
Catalog number 100120TS
also known as 100120
Price (INR)
-
Concentration:
1000 ng/mL

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Methadone Metabolite Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration1000 ng/mL
DescriptionDRI Methadone Metabolite Drugs of Abuse Calibrators
Packaging1 Bottle
Quantity1 x 10 mL
TypeCalibrator
Detectable AnalytesEDDP
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C