The Dynabeads Protein A and Magnet Starter Pack combines Dynabeads Protein A magnetic beads for immunoprecipitation with the magnet preferredRead more
Have Questions?
Change view
Catalog Number
Description
10013D
10016D
2 Options
Catalog number 10013D
Price (INR)
114,225.00
Each
Price (INR)
114,225.00
Each
The Dynabeads Protein A and Magnet Starter Pack combines Dynabeads Protein A magnetic beads for immunoprecipitation with the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience. Each mL of beads is sufficient for 20 reactions, and the magnet holds 16 x 1.5 mL microcentrifuge tubes. Dynabeads Protein A beads and the DynaMag-2 magnet are also available separately.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Ligand TypeProtein A
Quantity40 reactions
Shipping ConditionRoom Temperature
Product LineDynaMag, Dynabeads
TypeMagnet Starter Pack
Unit SizeEach
Contents & Storage
• 2 x 1 mL Dynabeads Protein A beads • 1 x DynaMag-2 magnet