Product Image
Thermo Scientific™
DRI™ Ecstasy Drugs of Abuse Assays
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100146813 x 18 mL
1000761 x 500 mL
Catalog number 10014681
Price (INR)
-
Quantity:
3 x 18 mL
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • One of the largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Select DAU Control Set
DescriptionDRI Ecstasy Drugs of Abuse Assay for Indiko
Quantity3 x 18 mL
Sample TypeUrine
TypeReagent
Detectable AnalytesMethylenedioxyamphetamine (MDA), Methylenedioxy-N-ethylamphetamine (MDEA), Methylenedioxymethamphetamine (MDMA)
DoA CalibratorsEcstasy Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C