Product Image
Thermo Scientific™
DRI™ Indiko™ Creatinine-Detect™ Specimen Validity Test
Rise to the challenge of combating the use of additives and adulterants to mask controlled substances with the Thermo Scientific™ DRI™ Indiko™ Creatinine-Detect™ kit.
Have Questions?
Catalog NumberQuantity
100156383 x 18 mL
Catalog number 10015638
Price (INR)
-
Quantity:
3 x 18 mL
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Indiko™ Creatinine-Detect™ Specimen Validity Tests. Liquid and ready-to-use, these tests can be performed on a variety of general chemistry analyzers.
  • Complete and automated way to test for sample integrity and adulteration
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionDRI Creatine-Detect Assay Specimen Validity Test
Quantity3 x 18 mL
Sample TypeUrine
Shelf LifeUntil Label Expiry Date
TypeReagent
Detectable AnalytesCreatinine
Unit SizeEach
Contents & Storage
2°C to 8°C