Save time in reagent preparation while maintaining high analytical accuracy with Thermo Scientific™ DRI™ Benzodiazepine Drugs of Abuse Assays.
Have Questions?
Catalog Number
Quantity
10015644
3 x 18 mL
Catalog number 10015644
HSN 38229090
Price (INR)
20,335.00
Each
Add to cart
Quantity:
3 x 18 mL
Price (INR)
20,335.00
Each
Add to cart
Save time in reagent preparation while maintaining high analytical accuracy with Thermo Scientific™ DRI™ Benzodiazepine Drugs of Abuse Assays. These liquid, ready-to-use assays offer applications for an array of clinical chemistry analyzers and have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
Qualitative or semi-quantitative options for results
Excellent correlation to GC/MS
Variety of calibrators and multiconstituent control sets
Specifications
Control SetsMGC Primary DAU Control Set
DescriptionDRI Benzodiazepine Drugs of Abuse Assay for Indiko