Product Image
Thermo Scientific™
DRI™ Indiko™ pH-Detect™ Specimen Validity Test
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Indiko™ pH-Detect™ Specimen Validity Tests.
Have Questions?
Catalog NumberQuantity
100156546 x 18 mL
Catalog number 10015654
Price (INR)
-
Quantity:
6 x 18 mL
Combat the use of additives and adulterants to mask controlled substances in urine samples with the Thermo Scientific™ DRI™ Indiko™ pH-Detect™ Specimen Validity Test. Liquid and ready-to-use, these tests can be performed on a variety of general chemistry analyzers.
The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution. The adulteration assays provide a complete and automated way to test for sample integrity and adulteration.
  • Complete and automated way to test for sample integrity and adulteration
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionDRI pH-Detect Assay Specimen Validity Test
Quantity6 x 18 mL
Sample TypeUrine
Shelf LifeUntil Label Expiry Date
TypeReagent
Detectable AnalytespH
Unit SizeEach
Contents & Storage
2°C to 8°C