Product Image
Thermo Scientific™
CEDIA™ Propoxyphene Drugs of Abuse Assays
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Propoxyphene Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
100170TS3 x 17 mL, TSC
1001711 x 65 mL, MCC
16615231 x 495 mL, LC
Catalog number 100170TS
Price (INR)
-
Quantity:
3 x 17 mL, TSC
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Propoxyphene Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • One of the largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Propoxyphene Drugs of Abuse Assays
Quantity3 x 17 mL, TSC
Sample TypeUrine
TypeReagent
Detectable AnalytesPropoxyphene (PPX)
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C