QMS™ Therapeutic Drug Monitoring (TDM) Assays
Actual product may vary
QMS™ Therapeutic Drug Monitoring (TDM) Assays
QMS™ Therapeutic Drug Monitoring (TDM) Assays
Thermo Scientific™

QMS™ Therapeutic Drug Monitoring (TDM) Assays

Utilize the precision, consistency and optimized performance of Thermo Scientific™ QMS™ TDM Assays, ready-to-use, liquid immunoassays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityDetectable Analytes
100171071 x 19 mL, 1 x 8 mLGentamicin
03739102 x 19 mL, 2 x 7 mLAmikacin
03746451 x 21 mL, 1 x 9 mLTeicoplanin
100143901 x 22 mL, 1 x 9 mLGentamicin
03737952 x 19 mLLamotrigine
03739362 x 19 mL, 2 x 7 mLQuinidine
03741401 x 22 mL, 1 x 16 mLTopiramate
03735892 x 22 mLVancomycin
03735712 x 22 mL, 2 x 8 mLZonisamide
100171091 x 14 mL, 1 x 12 mLTobramycin
Catalog number 10017107
Price (INR)
-
Quantity:
1 x 19 mL, 1 x 8 mL
Detectable Analytes:
Gentamicin
Accurately measure therapeutic drug levels in serum and plasma with the Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays. These liquid, ready-to-use assays are a convenient alternative to more expensive and time-consuming technologies. QMS (Quantitative Microsphere System) immunoassays have excellent precision and accuracy, optimized performance on a wide range of general chemistry analyzers and lot-to-lot consistency.
Reduces Labor
  • Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
  • Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
Ready-to-use
  • Liquid stable and ready-to-use
  • Applicable to general chemistry analyzers
  • Validated parameters assure lab-to-lab reproducibility
  • Superb reagent stability
Optimal Sensitivity
  • Excellent correlation to reference methods
  • Optimal sensitivity, precision and dynamic range
  • Low cross-reactivity with metabolites and commonly co-administered drugs
  • Low interference from endogenous substances
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionGentamicin Assay for Indiko
Quantity1 x 19 mL, 1 x 8 mL
Sample TypePlasma, Serum
TypeReagent
Detectable AnalytesGentamicin
Unit SizeEach
Contents & Storage
2°C to 8°C