Product Image
Thermo Scientific™

CEDIA™ Phencyclidine (PCP) Drugs of Abuse Assays

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Phencyclidine (PCP) Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1001731 x 65 mL, MCC
1001723 x 17 mL, TSC
18157841 x 495 mL, LC
Catalog number 100173
Price (INR)
-
Quantity:
1 x 65 mL, MCC
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of CEDIA™ Phencyclidine (PCP) Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionCEDIA Phencycidine (PCP) Assay
Quantity1 x 65 mL, MCC
Sample TypeUrine
TypeReagent
Detectable AnalytesPhencyclidine (PCP)
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C